3-Chlor-2-fluorbenzamid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chlor-2-fluorbenzoesäureamid

PC0031-25G PC0031-10G PC0031-1G
APOSPC0031-25GEA 75.4 EUR
APOSPC0031-25G APOSPC0031-10G APOSPC0031-1G
3-Chlor-2-fluorbenzamid 97%
3-Chlor-2-fluorbenzamid
Formel: C₇H₅ClFNO
Molecular Weight: 173.57 g/mol
Schmelzpunkt: 125…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042205
CAS-Nummer: 104326-94-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR