5-Chlor-2-fluorbenzamid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chlor-2-fluorbenzoesäureamid, 3-Chlor-6-fluorbenzoesäureamid
PC0033-25G
PC0033-5G
APOSPC0033-25GEA
125
EUR
APOSPC0033-25G
APOSPC0033-5G
5-Chlor-2-fluorbenzamid 97%
5-Chlor-2-fluorbenzamid
Formel:
C₇H₅ClFNO Schmelzpunkt: 134…137 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631435 CAS-Nummer: 261762-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...