5-Chlor-2-fluorbenzonitril 97%

Lieferant: Apollo Scientific
Gefahr

PC0037-100G PC0037-500G PC0037-50G PC0037-25G
APOSPC0037-100GEA 97.3 EUR
APOSPC0037-100G APOSPC0037-500G APOSPC0037-50G APOSPC0037-25G
5-Chlor-2-fluorbenzonitril 97%
5-Chlor-2-fluorbenzonitril
Formel: C₇H₃ClFN
Molecular Weight: 155,56 g/mol
Schmelzpunkt: 67…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631434
CAS-Nummer: 57381-34-7
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR