3-(3-Fluor-4-methoxybenzoyl)propionsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC0039-10G
PC0039-1G
PC0039-5G
APOSPC0039-10GEA
63.8
EUR
APOSPC0039-10G
APOSPC0039-1G
APOSPC0039-5G
3-(3-Fluor-4-methoxybenzoyl)propionsäure 97%
3-(3-Fluor-4-methoxybenzoyl)propionsäure
Formel:
C₁₁H₁₁FO₄ Molecular Weight: 226.20 g/mol Schmelzpunkt: 169…173 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020538 CAS-Nummer: 347-63-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...