5-[5-(Trifluoromethyl)isoxazol-3-yl]thiophene-2-sulphonyl chloride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC0042-1G
PC0042-2.5G
PC0042-5G
APOSPC0042-1GEA
69.6
EUR
APOSPC0042-1G
APOSPC0042-2.5G
APOSPC0042-5G
5-[5-(Trifluoromethyl)isoxazol-3-yl]thiophene-2-sulphonyl chloride 97%
5-[5-(Trifluoromethyl)isoxazol-3-yl]thiophene-2-sulphonyl chloride
Formel:
C₈H₃ClF₃NO₃S₂ Molecular Weight: 317.69 g/mol Schmelzpunkt: 106…111 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00831001 CAS-Nummer: 229956-98-3 UN: 1759 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...