2-Chlor-3-ethylbenzoxazoliumtetrafluoroborat 97%

Lieferant: Apollo Scientific

PC0062-25G PC0062-5G
APOSPC0062-25GEA 364 EUR
APOSPC0062-25G APOSPC0062-5G
2-Chlor-3-ethylbenzoxazoliumtetrafluoroborat 97%
2-Chloro-3-ethylbenzoxazolium tetrafluoroborate

Mild reagent which is able to activate alcohols to nucleophiles: Chem Lett., 383 (1997). Can act as a gentle reducing agent in the presence of base.

Formel: C₉H₉BClF₄NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00011943
CAS-Nummer: 63212-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR