3-Chlor-2-fluorbenzylamin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(3-Chloro-2-fluorophenyl)methanamine
PC0064-10G
PC0064-25G
PC0064-5G
APOSPC0064-10GEA
66.7
EUR
APOSPC0064-10G
APOSPC0064-25G
APOSPC0064-5G
3-Chlor-2-fluorbenzylamin 98%
3-Chlor-2-fluorbenzylamin
Formel:
C₇H₇ClFN Molecular Weight: 159,59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631391 CAS-Nummer: 72235-55-3 UN: 2735 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...