3-Chlor-5-fluorbenzaldehyd 97%

Lieferant: Apollo Scientific
Achtung

PC0067-1G PC0067-25G PC0067-5G
APOSPC0067-1GEA 21.9 EUR
APOSPC0067-1G APOSPC0067-25G APOSPC0067-5G
3-Chlor-5-fluorbenzaldehyd 97%
3-Chloro-5-fluorobenzaldehyde
Formel: C₇H₄ClFO
Molecular Weight: 158.56 g/mol
Schmelzpunkt: 38…42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02261734
CAS-Nummer: 90390-49-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR