5-Chlor-2-fluorbenzylamin

Lieferant: Apollo Scientific
Gefahr

PC0068-250MG PC0068-5G PC0068-1G
APOSPC0068-250MGEA 21.9 EUR
APOSPC0068-250MG APOSPC0068-5G APOSPC0068-1G
5-Chlor-2-fluorbenzylamin
5-Chlor-2-fluorbenzylamin
Formel: C₇H₇ClFN
Lagertemperatur: Raumtemperatur
MDL: MFCD01631440
CAS-Nummer: 261723-26-6
UN: 2735
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR