2-Fluorphenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Fluorphenyl)metaborsäure, 2-Fluorbenzolboronsäure, Dihydroxy(2-fluorphenyl)boran
PC0073-100G
PC0073-25G
PC0073-500G
APOSPC0073-100GEA
119
EUR
APOSPC0073-100G
APOSPC0073-25G
APOSPC0073-500G
2-Fluorphenylboronsäure 97%
2-Fluorphenylboronsäure
Formel:
C₆H₆BFO₂ Molecular Weight: 139,92 g/mol Schmelzpunkt: 101…118 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00674013 CAS-Nummer: 1993-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...