2'-Chlor-6'-fluor-3'-methylacetophenon

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Chloro-6-fluoro-3-methylphenyl)ethanone

PC0085-25G PC0085-5G PC0085-100G
APOSPC0085-25GEA 168 EUR
APOSPC0085-25G APOSPC0085-5G APOSPC0085-100G
2'-Chlor-6'-fluor-3'-methylacetophenon
2'-Chlor-6'-fluor-3'-methylacetophenon
Formel: C₉H₈ClFO
Molecular Weight: 186,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631368
CAS-Nummer: 261762-63-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR