2-Chlor-4'-fluorpropiophenon 98%
Lieferant: Apollo Scientific
PC0087-1G
PC0087-5G
APOSPC0087-1GEA
109
EUR
APOSPC0087-1G
APOSPC0087-5G
2-Chlor-4'-fluorpropiophenon 98%
2-Chlor-4'-fluorpropiophenon
Formel:
C₉H₈ClFO Siedepunkt: 71 °C/1mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060275 CAS-Nummer: 81112-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...