2-Chlor-6-fluor-3-methylbenzaldehyd 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chlor-6-fluor-m-tolylaldehyd, 2-Chlor-6-fluor-m-methylbenzaldehyd, 2-Chlor-6-fluor-3-tolylaldehyd, 2-Chlor-6-fluor-m-tolualdehyd, 2-Chlor-6-fluor-3-tolualdehyd
PC0093-25G
PC0093-5G
PC0093-1G
APOSPC0093-25GEA
203
EUR
APOSPC0093-25G
APOSPC0093-5G
APOSPC0093-1G
2-Chlor-6-fluor-3-methylbenzaldehyd 95%
2-Chloro-6-fluoro-3-methylbenzaldehyde
Formel:
C₈H₆ClFO Molecular Weight: 172.59 Schmelzpunkt: 26…33 °C Flammpunkt: 110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631360 CAS-Nummer: 104451-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...