6-Chlor-2-fluor-3-methylbenzamid 97%
Lieferant: Apollo Scientific
PC0096-25G
PC0096-5G
PC0096-1G
APOSPC0096-25GEA
372
EUR
APOSPC0096-25G
APOSPC0096-5G
APOSPC0096-1G
6-Chlor-2-fluor-3-methylbenzamid 97%
6-Chlor-2-fluor-3-methylbenzamid
Formel:
C₈H₇ClFNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631462 CAS-Nummer: 286474-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...