2-Chlor-6-fluor-m-toluylsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-6-fluoro-3-methylbenzoic acid

PC0097-5G PC0097-1G PC0097-25G
APOSPC0097-5GEA 55.1 EUR
APOSPC0097-5G APOSPC0097-1G APOSPC0097-25G
2-Chlor-6-fluor-m-toluylsäure
2-Chlor-6-fluor-m-toluylsäure
Formel: C₈H₆ClFO₂
Molecular Weight: 188,59 g/mol
Schmelzpunkt: 116…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631359
CAS-Nummer: 32890-89-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR