6-Chlor-2-fluor-m-toluylsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Chloro-2-fluoro-3-methylbenzoic acid
PC0099-25G
PC0099-5G
PC0099-100G
APOSPC0099-25GEA
418
EUR
APOSPC0099-25G
APOSPC0099-5G
APOSPC0099-100G
6-Chlor-2-fluor-m-toluylsäure 98%
6-Chlor-2-fluor-m-toluylsäure
Formel:
C₈H₆ClFO₂ Molecular Weight: 188,59 g/mol Schmelzpunkt: 135…136 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631369 CAS-Nummer: 32890-90-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...