2-Chloro-6-fluoro-3-methylbenzoyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Chloro-6-fluoro-3-methylbenzene-1-carbonyl chloride, 2-Chloro-6-fluoro-3-methylbenzoylchloride, 2-Chloro-6-fluoro-3-toluoyl chloride

PC0100-1G PC0100-25G PC0100-5G
APOSPC0100-1GEA 40.6 EUR
APOSPC0100-1G APOSPC0100-25G APOSPC0100-5G
2-Chloro-6-fluoro-3-methylbenzoyl chloride
2-Chloro-6-fluoro-3-methylbenzoyl chloride
Formel: C₈H₅Cl₂FO
Molecular Weight: 207,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631371
CAS-Nummer: 261762-82-7
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR