6-Chlor-2-fluor-3-toluoylchlorid 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 6-Chlor-2-fluor-3-methylbenzoylchlorid, 6-Chlor-2-fluor-3-methylbenzoesäurechlorid, 6-Chlor-2-fluor-3-methylbenzol-1-carbonylchlorid, 6-Chlor-2-fluor-3-tolylsäurechlorid

PC0103-1G PC0103-5G PC0103-25G
APOSPC0103-1GEA 31.9 EUR
APOSPC0103-1G APOSPC0103-5G APOSPC0103-25G
6-Chlor-2-fluor-3-toluoylchlorid 95%
6-Chlor-2-fluor-3-toluoylchlorid
Formel: C₈H₅Cl₂FO
Molecular Weight: 207,03 g/mol
Siedepunkt: 110 °C/1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01631361
CAS-Nummer: 261762-81-6
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR