2-Chloro-6-fluoro-3-methylbenzyl alcohol 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC0108-1G
PC0108-5G
PC0108-10G
APOSPC0108-1GEA
34.8
EUR
APOSPC0108-1G
APOSPC0108-5G
APOSPC0108-10G
2-Chloro-6-fluoro-3-methylbenzyl alcohol 98%
2-Chloro-6-fluoro-3-methylbenzyl alcohol
Formel:
C₈H₈ClFO Molecular Weight: 174.6 g/mol Schmelzpunkt: 56…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631362 CAS-Nummer: 261762-83-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...