2-Quinolin-2-yl-1-(4-trifluoromethylphenyl)ethanol 95+%
Lieferant: Apollo Scientific
Synonyme:
2-(Quinolin-2-yl)-1-(4-(trifluoromethyl)phenyl)ethanol
PC0110-10G
PC0110-5G
PC0110-1G
APOSPC0110-10GEA
606
EUR
APOSPC0110-10G
APOSPC0110-5G
APOSPC0110-1G
2-Quinolin-2-yl-1-(4-trifluoromethylphenyl)ethanol 95+%
2-Quinolin-2-yl-1-(4-trifluoromethylphenyl)ethanol
Formel:
C₁₈H₁₄F₃NO Molecular Weight: 317.31 g/mol Schmelzpunkt: 171…173 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424493 CAS-Nummer: 496947-30-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...