(6-Chlor-2-fluor-3-methylphenyl)essigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC0122-1G
PC0122-5G
PC0122-25G
APOSPC0122-1GEA
71.2
EUR
APOSPC0122-1G
APOSPC0122-5G
APOSPC0122-25G
(6-Chlor-2-fluor-3-methylphenyl)essigsäure 98%
(6-Chlor-2-fluor-3-methylphenyl)essigsäure
Formel:
C₉H₈ClFO₂ Molecular Weight: 202.61 g/mol Schmelzpunkt: 130…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631375 CAS-Nummer: 261762-93-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...