6-Chloro-2-fluoro-3-methylphenylacetonitrile 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC0127-1G
PC0127-25G
PC0127-5G
APOSPC0127-1GEA
105
EUR
APOSPC0127-1G
APOSPC0127-25G
APOSPC0127-5G
6-Chloro-2-fluoro-3-methylphenylacetonitrile 98%
6-Chloro-2-fluoro-3-methylphenylacetonitrile
Formel:
C₉H₇ClFN Molecular Weight: 183.61 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631374 CAS-Nummer: 261762-95-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...