(3-Chlor-4-fluorphenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-4-fluorophenylacetic acid

PC0134-5G PC0134-1G
APOSPC0134-5GEA 65.4 EUR
APOSPC0134-5G APOSPC0134-1G
(3-Chlor-4-fluorphenyl)essigsäure 98%
(3-Chlor-4-fluorphenyl)essigsäure
Formel: (C₆H₃ClF)CH₂COOH
Molecular Weight: 188,59 g/mol
Schmelzpunkt: 55…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631555
CAS-Nummer: 705-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR