(5-Chlor-2-fluorphenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(5-Chloro-2-fluorophenyl)acetic acid

PC0135-10G PC0135-5G PC0135-1G
APOSPC0135-10GEA 45.1 EUR
APOSPC0135-10G APOSPC0135-5G APOSPC0135-1G
(5-Chlor-2-fluorphenyl)essigsäure 98%
(5-Chlor-2-fluorphenyl)essigsäure
Formel: (C₆H₃ClF)CH₂COOH
Molecular Weight: 188,59 g/mol
Schmelzpunkt: 116…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631439
CAS-Nummer: 261762-97-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR