3-Chlor-4-fluorphenylacetonitril

Lieferant: Apollo Scientific
Gefahr

PC0137-100G PC0137-25G PC0137-5G
APOSPC0137-100GEA 336 EUR
APOSPC0137-100G APOSPC0137-25G APOSPC0137-5G
3-Chlor-4-fluorphenylacetonitril
3-Chlor-4-fluorphenylacetonitril
Formel: C₈H₅ClFN
Molecular Weight: 169,5834 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631553
CAS-Nummer: 658-98-0
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR