3-Chloro-4-fluoro-DL-phenylglycine

Lieferant: Apollo Scientific
Achtung

PC0139-10G PC0139-5G PC0139-1G
APOSPC0139-10GEA 0 EUR
APOSPC0139-10G APOSPC0139-5G APOSPC0139-1G
3-Chloro-4-fluoro-DL-phenylglycine
3-Chloro-4-fluoro-DL-phenylglycine
Formel: C₈H₇ClFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01631397
CAS-Nummer: 261762-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR