2-Fluor-3-hydroxypyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluor-3-pyridinol
PC01476-25G
PC01476-5G
PC01476-100G
APOSPC01476-25GEA
58
EUR
APOSPC01476-25G
APOSPC01476-5G
APOSPC01476-100G
2-Fluor-3-hydroxypyridin
2-Fluor-3-hydroxypyridin
Formel:
C₅H₄FNO Molecular Weight: 113.09 g/mol Schmelzpunkt: 131…133 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04112569 CAS-Nummer: 174669-74-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...