3-Bromo-4-fluorobenzenesulphonyl chloride 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Bromo-4-fluorobenzene-1-sulphonyl chloride
PC01491-10G
PC01491-1G
PC01491-5G
APOSPC01491-10GEA
412
EUR
APOSPC01491-10G
APOSPC01491-1G
APOSPC01491-5G
3-Bromo-4-fluorobenzenesulphonyl chloride 98%
3-Bromo-4-fluorobenzenesulphonyl chloride
Formel:
C₆H₃BrClFO₂S Molecular Weight: 273,51 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185363 CAS-Nummer: 631912-19-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...