3-Chlor-2-fluor-5-(trifluormethyl)benzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC0150-100G
PC0150-25G
PC0150-5G
APOSPC0150-100GEA
719
EUR
APOSPC0150-100G
APOSPC0150-25G
APOSPC0150-5G
3-Chlor-2-fluor-5-(trifluormethyl)benzoesäure 98%
3-Chloro-2-fluoro-5-(trifluoromethyl)benzoic acid
Formel:
C₈H₃ClF₄O₂ Molecular Weight: 242.56 g/mol Schmelzpunkt: 89…91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631509 CAS-Nummer: 129931-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...