5-Brom-2-(trifluormethyl)anilin 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-4-bromobenzotrifluoride
PC01503-100G
PC01503-25G
PC01503-5G
APOSPC01503-100GEA
557
EUR
APOSPC01503-100G
APOSPC01503-25G
APOSPC01503-5G
5-Brom-2-(trifluormethyl)anilin 95%
5-Brom-2-(trifluormethyl)anilin
Formel:
C₇H₅BrF₃N Molecular Weight: 240.0200 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082853 CAS-Nummer: 703-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...