4-Brom-2-fluorthioanisol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Bromo-2-fluorophenyl)(methyl)sulphane
PC01510-1G
PC01510-5G
PC01510-25G
APOSPC01510-1GEA
52.2
EUR
APOSPC01510-1G
APOSPC01510-5G
APOSPC01510-25G
4-Brom-2-fluorthioanisol 95%
4-Brom-2-fluorthioanisol
Formel:
C₇H₆BrFS Molecular Weight: 221,0899 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12756455 CAS-Nummer: 221030-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...