4-Brom-2-fluorthiophenol
Lieferant: Apollo Scientific
|
Achtung
|
PC01514-1G
PC01514-5G
PC01514-25G
APOSPC01514-1GEA
53.8
EUR
APOSPC01514-1G
APOSPC01514-5G
APOSPC01514-25G
4-Brom-2-fluorthiophenol
4-Brom-2-fluorthiophenol
Formel:
C₆H₄BrFS Lagertemperatur: Kühlschrank |
MDL:
MFCD09475475 CAS-Nummer: 174414-93-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...