3-Chlor-2-fluor-5-(trifluormethyl)benzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

PC0154-10G PC0154-5G PC0154-1G
APOSPC0154-10GEA 197 EUR
APOSPC0154-10G APOSPC0154-5G APOSPC0154-1G
3-Chlor-2-fluor-5-(trifluormethyl)benzylalkohol 98%
3-Chlor-2-fluor-5-(trifluormethyl)benzylalkohol
Formel: C₈H₅ClF₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD01631512
CAS-Nummer: 261763-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR