4-Fluor-2-nitrobenzoesäure 97%

Lieferant: Apollo Scientific
Achtung

PC0164-100G PC0164-25G
APOSPC0164-100GEA 81.2 EUR
APOSPC0164-100G APOSPC0164-25G
4-Fluor-2-nitrobenzoesäure 97%
4-Fluoro-2-nitrobenzoic acid
Formel: C₇H₄FNO₄
Molecular Weight: 185,11 g/mol
Schmelzpunkt: 140…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00024300
CAS-Nummer: 394-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR