4-Chloro-2-(trifluoromethyl)pyridine 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-(2-trifluoromethyl)pyridine

PC01677-25G PC01677-5G PC01677-1G
APOSPC01677-25GEA 155 EUR
APOSPC01677-25G APOSPC01677-5G APOSPC01677-1G
4-Chloro-2-(trifluoromethyl)pyridine 99%
4-Chloro-2-(trifluoromethyl)pyridine
Formel: C₆H₃ClF₃N
Molecular Weight: 181,542 g/mol
Siedepunkt: 137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368047
CAS-Nummer: 131748-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR