3-Fluor-2-iodanilin

Lieferant: Apollo Scientific
Achtung

PC01678-1G PC01678-250MG PC01678-5G
APOSPC01678-1GEA 102 EUR
APOSPC01678-1G APOSPC01678-250MG APOSPC01678-5G
3-Fluor-2-iodanilin
3-Fluor-2-iodanilin
Formel: C₆H₅FIN
Molecular Weight: 237,0100 g/mol
Schmelzpunkt: 52…54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08458368
CAS-Nummer: 706752-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR