3-Chlor-2-fluor-5-(trifluormethyl)phenylacetonitril 98%
Lieferant: Apollo Scientific
PC0168-1G
PC0168-25G
PC0168-5G
APOSPC0168-1GEA
97.3
EUR
APOSPC0168-1G
APOSPC0168-25G
APOSPC0168-5G
3-Chlor-2-fluor-5-(trifluormethyl)phenylacetonitril 98%
3-Chlor-2-fluor-5-(trifluormethyl)phenylacetonitril
Formel:
C₉H₄ClF₄N Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631515 CAS-Nummer: 261763-15-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...