1-Chloro-3,3,3-trifluoroacetone
Lieferant: Apollo Scientific
|
Achtung
|
PC0171-25G
PC0171-5G
APOSPC0171-25GEA
571
EUR
APOSPC0171-25G
APOSPC0171-5G
1-Chloro-3,3,3-trifluoroacetone
1-Chloro-3,3,3-trifluoroacetone
Formel:
C₃H₂ClF₃O Molecular Weight: 146,5 g/mol Siedepunkt: 71…72 °C Dichte: 1,45 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631508 CAS-Nummer: 431-37-8 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...