1-Brom-1-chlor-2,2-difluorethylen 97%
Lieferant: Apollo Scientific
Synonyme:
1-Bromo-1-chlorodifluoroethylene
PC0173-1G
PC0173-5G
APOSPC0173-1GEA
245
EUR
APOSPC0173-1G
APOSPC0173-5G
1-Brom-1-chlor-2,2-difluorethylen 97%
1-Brom-1-chlor-2,2-difluorethylen
Formel:
C₂BrClF₂ Molecular Weight: 177,38 g/mol Siedepunkt: 40…42 °C Dichte: 1,888 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00166977 CAS-Nummer: 758-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...