3-Chloro-4-(trifluoromethoxy)benzaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-4-trifluoromethoxybenzaldehyde
PC0174-1G
PC0174-25G
PC0174-5G
APOSPC0174-1GEA
21.9
EUR
APOSPC0174-1G
APOSPC0174-25G
APOSPC0174-5G
3-Chloro-4-(trifluoromethoxy)benzaldehyde
3-Chloro-4-(trifluoromethoxy)benzaldehyde
Formel:
C8H4ClF3O2 Molecular Weight: 224,5600 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631561 CAS-Nummer: 83279-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...