4-Chlor-3-(trifluormethyl)benzylalkohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Chloro-3-(trifluoromethyl)phenyl)methanol

PC0192-1G PC0192-25G PC0192-5G
APOSPC0192-1GEA 21.9 EUR
APOSPC0192-1G APOSPC0192-25G APOSPC0192-5G
4-Chlor-3-(trifluormethyl)benzylalkohol
4-Chlor-3-(trifluormethyl)benzylalkohol
Formel: C₈H₆ClF₃O
Molecular Weight: 210,5809 g/mol
Schmelzpunkt: 44…46 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631538
CAS-Nummer: 65735-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR