(2-Chlor-5-(trifluormethyl)phenyl)essigsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid

PC0199-1G PC0199-5G PC0199-10G
APOSPC0199-1GEA 33.5 EUR
APOSPC0199-1G APOSPC0199-5G APOSPC0199-10G
(2-Chlor-5-(trifluormethyl)phenyl)essigsäure
(2-Chlor-5-(trifluormethyl)phenyl)essigsäure
Formel: C₉H₆ClF₃O₂
Molecular Weight: 238,59 g/mol
Schmelzpunkt: 112…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631471
CAS-Nummer: 22893-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR