(5-Chlor-2-(trifluormethyl)phenyl)essigsäure 98%
Lieferant: Apollo Scientific
PC0200-1G
PC0200-5G
APOSPC0200-1GEA
120
EUR
APOSPC0200-1G
APOSPC0200-5G
(5-Chlor-2-(trifluormethyl)phenyl)essigsäure 98%
(5-Chlor-2-(trifluormethyl)phenyl)essigsäure
Formel:
C₉H₆ClF₃O₂ Molecular Weight: 238,59 g/mol Schmelzpunkt: 117…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631358 CAS-Nummer: 261763-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...