α-Brom-2,4-dichlor-5-fluortoluol 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,4-Dichlor-5-fluorbenzylbromid

PC0212-250MG PC0212-1G PC0212-5G
APOSPC0212-250MGEA 21.9 EUR
APOSPC0212-250MG APOSPC0212-1G APOSPC0212-5G
α-Brom-2,4-dichlor-5-fluortoluol 98%
α-Brom-2,4-dichlor-5-fluortoluol
Formel: C₇H₄BrCl₂F
Molecular Weight: 257.92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631381
CAS-Nummer: 261763-27-3
UN: 1760
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR