2,2-Difluor-1,3-benzodioxol-4-carbonylchlorid

Lieferant: Apollo Scientific
Gefahr

PC0220-1G PC0220-5G
APOSPC0220-1GEA 222 EUR
APOSPC0220-1G APOSPC0220-5G
2,2-Difluor-1,3-benzodioxol-4-carbonylchlorid
2,2-Difluor-1,3-benzodioxol-4-carbonylchlorid
Formel: C₈H₃ClF₂O₃
Lagertemperatur: Gefrierschrank
MDL: MFCD01631475
CAS-Nummer: 127163-51-3
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR