2',3'-Difluor-4'-methylacetophenon 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2,3-Difluoro-4-methylphenyl)ethanone
PC0228-10G
PC0228-1G
PC0228-5G
APOSPC0228-10GEA
209
EUR
APOSPC0228-10G
APOSPC0228-1G
APOSPC0228-5G
2',3'-Difluor-4'-methylacetophenon 99%
2',3'-Difluor-4'-methylacetophenon
Formel:
C₉H₈F₂O Molecular Weight: 170,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631650 CAS-Nummer: 261763-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...