2,3-Difluor-4-methylanisol 98%

Lieferant: Apollo Scientific

PC0231-10G PC0231-5G PC0231-1G
APOSPC0231-10GEA 134 EUR
APOSPC0231-10G APOSPC0231-5G APOSPC0231-1G
2,3-Difluor-4-methylanisol 98%
2,3-Difluor-4-methylanisol
Formel: C₈H₈F₂O
Molecular Weight: 158.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631321
CAS-Nummer: 261763-32-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR