2,3-Difluor-4-methylbenzylbromid
Lieferant: Apollo Scientific
|
Gefahr
|
PC0257-10G
PC0257-5G
APOSPC0257-10GEA
412
EUR
APOSPC0257-10G
APOSPC0257-5G
2,3-Difluor-4-methylbenzylbromid
2,3-Difluor-4-methylbenzylbromid
Formel:
C₈H₇BrF₂ Molecular Weight: 221.04 g/mol Siedepunkt: 78 °C/1mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631647 CAS-Nummer: 261763-43-3 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...