2,6-Difluor-3-methylbenzylbromid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-(Bromomethyl)-1,3-difluoro-4-methylbenzene

PC0258-10G PC0258-5G
APOSPC0258-10GEA 219 EUR
APOSPC0258-10G APOSPC0258-5G
2,6-Difluor-3-methylbenzylbromid
2,6-Difluor-3-methylbenzylbromid
Formel: C₈H₇BrF₂
Molecular Weight: 221,04 g/mol
Siedepunkt: 76 °C/3mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01631336
CAS-Nummer: 261763-44-4
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR