(2,6-Difluor-3-methylphenyl)acetonitril

Lieferant: Apollo Scientific
Gefahr

PC0265-1G PC0265-5G PC0265-25G
APOSPC0265-1GEA 0 EUR
APOSPC0265-1G APOSPC0265-5G APOSPC0265-25G
(2,6-Difluor-3-methylphenyl)acetonitril
(2,6-Difluor-3-methylphenyl)acetonitril
Formel: C₉H₇F₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD01631337
CAS-Nummer: 261944-04-1
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR